Cole-Parmer Tuff-Tip In-Line pH Probe, DJ/CPVC/No ATC; /BNC
Representative image only.
Cole-Parmer Item # EW-27003-01
Less chance of breakage
  • Feature a 10-ft cable with BNC connector
  • 1 year warranty
PRODUCT OPTIONS
$158.00USD / Each
BUY MORE & SAVE
  • 1-5$158.00USD / Each
  • 6-11$142.20USD / Each
  • 12-23$134.30USD / Each
  • 24+$126.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeDouble junction
  • Electrode Body MaterialCPVC
  • Min pH (pH)12
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • Max Pressure (PSI)100
  • Thread Size NPT(M) (in)3/4
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionTuff-Tip In-Line pH Probe, DJ/CPVC/No ATC; /BNC
  • Warranty1 year
1 Year WarrantyCole-Parmer Exclusive
Key Features
  • Feature a 10-ft cable with BNC connector
  • 1 year warranty
More About this Item
pH electrodes are ideal for low temperatures, nonaqueous solutions, and pH measurements from 0 to 12.