This item is now discontinued. We recommend the following as suggested replacements: EW-36620-55
Cole-Parmer StableTemp Single Block for Conical Bottom Centrifuge Tubes, 15 Ml
Representative image only.
Cole-Parmer StableTemp Item # EW-36600-28
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Block Number Of Openings12
  • Block - Tube / Vial Size Accepted (mL)15
  • Height (in)2
  • Width (in)3
  • Length (in)3 3/4
  • DescriptionSingle Block for Conical Bottom Centrifuge Tubes, 15 Ml
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Modular blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature. Centrifuge Tube blocks are single size blocks and are available in 15mL or 50mL tube sizes.
15mL centrifuge tube block. Conical bottom. Single block