Cole-Parmer StableTemp Double Block for PCR, 96 Well PCR Plate
Representative image only.
Cole-Parmer StableTemp Item # EW-36600-51
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Block Number Of Openings1
  • Combination Block -Tube / Vial Size Accepted96-well PCR plate
  • Height (in)2 1/4
  • Width (in)3 3/4
  • Length (in)6
  • DescriptionDouble Block for PCR, 96 Well PCR Plate
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Modular blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block walls allow for maximum heat retention. The PCR Plate block has a thermometer well for measuring block temperature and is a double size block. Please note that the PCR Tube and PCR Strip blocks do not have a thermometer well and are single blocks. Tapered tube wells for 0.2mL tubes are spaced for easy access and removal.