Cole-Parmer SS, 1/4" NPT (f) X 1/4" NPT (f), VAC To 500 PSI
Representative image only.
Cole-Parmer Item # EW-68340-18
Protect any gauge with a NPT(M) connector from contaminated, corrosive, hot, or viscous media
  • Protect transducers, switches, and gauges 3 1/2'' in diameter or smaller
  • Fill/bleed connection is standard
  • 1 year warranty
$121.55USD / Each
$143.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure (PSI)500
  • Process Connection1/4\" NPT(F)
  • Instrument Connection Npt(F) (in)1/4
  • DescriptionSS, 1/4\" NPT (f) X 1/4\" NPT (f), VAC To 500 PSI
  • Warranty1 year
1 Year Warranty
Key Features
  • Protect transducers, switches, and gauges 3 1/2'' in diameter or smaller
  • Fill/bleed connection is standard
  • 1 year warranty
More About this Item
Gauge Guards are used to isolate and protect pressure or vacuum instruments used on ultra-pure or highly-corrosive fluid lines, whenever such fluids will corrode metal instrument components. Gauge Guards also protect against clogging of instruments by suspended solids or highly viscous fluids. Furthermore, they eliminate dead pocket areas within instruments where chemicals or organic matter could decompose. Gauge guards are a very cost-effective way to protect your very expensive transducers, gauges, or switches.
Process connections are 1/4'' and 1/2'' NPT (F), and 1 1/2'' and 2'' sanitary Tri-Clamp.