Cole-Parmer Essentials Solid Color-Coded Silicone Stoppers, Standard Size 2, Pink; 20/Bag
Representative image only.
Cole-Parmer Essentials Item # EW-06298-06
  • 6 month warranty
PRODUCT OPTIONS
$66.70USD / Bag of 20
BUY MORE & SAVE
  • 1-5$66.70USD / Bag of 20
  • 6-29$60.03USD / Bag of 20
  • 30-119$56.70USD / Bag of 20
  • 120+$53.36USD / Bag of 20
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number2
  • Top Outside Diameter (OD) (mm)20
  • Bottom Outside Diameter (OD) (mm)15.5
  • Height (mm)27
  • ColorPink
  • MaterialSilicone
  • DescriptionSolid Color-Coded Silicone Stoppers, Standard Size 2, Pink; 20/Bag
  • Warranty6 month
6 Month Warranty
Key Features
  • 6 month warranty
More About this Item
Silicone bubbles make these stoppers lightweight—they float on water. Withstand temperatures from –76 to 392°>F (–60 to 200°>C). Stoppers are available in 14 color-coded sizes, given in both standard and Japanese size numbers.
Softness and flexibility allow stoppers to fit a range of holes. Easily make openings for glassware fit your exact specifications with the optional cork borer set. 2 bg/cs.*