Cole-Parmer Sealing Mat for 96-Well Plates, 0.5- and 1.2 mL,TPE; 50/pk
Representative image only.
Cole-Parmer Item # EW-67104-69
Prevent spillage and contamination between wells
  • Tight seal reduces evaporation losses
  • Easily pierceable with pipette tips
  • Free of RNase, DNase, and endotoxins
  • Suitable for 96-well deep well plates
PRODUCT OPTIONS
$340.50USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • MaterialTPE
  • DescriptionSealing Mat for 96-Well Plates, 0.5- and 1.2 mL; 50/pk
Key Features
  • Tight seal reduces evaporation losses
  • Easily pierceable with pipette tips
  • Free of RNase, DNase, and endotoxins
  • Suitable for 96-well deep well plates
More About this Item
Choose from TPE or EVA materials to meet your application needs.