Cole-Parmer Essentials PP Wall Mount Drying Rack with Pegs, 20 Place

Representative image only.
Cole-Parmer Essentials Item # EW-06045-91
Don’t let water collect
  • Draining port allows water to drain back to sink Pegs screw-in for removal to accommodate different size labware Features 5 tiers; 20 pegs
  • 1 year warranty

$146.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Tubes Held20
  • Length (in)13 1/4
  • Width (in)2 1/4
  • Height (in)19 1/4
  • Length (mm)349
  • Width (mm)87
  • MaterialPolypropylene(PP)
  • DescriptionPP Wall Mount Drying Rack with Pegs, 20 Place
  • Warranty1 year
1 Year Warranty

Key Features

  • Draining port allows water to drain back to sink Pegs screw-in for removal to accommodate different size labware Features 5 tiers; 20 pegs
  • 1 year warranty

More About this Item

Polypropylene draining rack provides an excellent option for post-wash draining of glassware. Labware dries quickly on screw-in pegs. Rack features 5 tiers of 20 pegs, hooks for wall mounting, a wastewater tray, and draining port.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat