Cole-Parmer Essentials Nylon Membrane Filters; 0.2µm Pore, 25mm Dia, 100/Pk
Representative image only.
Cole-Parmer Essentials Item # EW-15945-24
Filter aqueous and organic solvents without prewetting
  • Use for HPLC sample preparation, to sterilize biological solutions/buffers, and vacuum degassing
PRODUCT OPTIONS
$21.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialNylon
  • Pore Size (µm)0.2
  • Diameter (mm)25
  • DescriptionNylon Membrane Filters; 0.2µm Pore, 25mm Dia, 100/Pk
ISO9001 Certified Supplier
Key Features
  • Use for HPLC sample preparation, to sterilize biological solutions/buffers, and vacuum degassing
More About this Item
Nonsterile nylon membranes are inherently hydrophilic and are compatible with many aqueous and organic solvents and solutions making them suitable for HPLC sample preparation, sterilizing biological solutions or buffers, and vacuum degassing. Membranes may be exposed to temperatures up to 75°C (167°F), and can withstand up to five autoclave cycles at 121°C for 30 minutes. Additional variations are available; call us for details.