Cole-Parmer NIST-Traceable Viscosity Standard, S60; 500 mL
Representative image only.
Cole-Parmer Item # EW-98944-76
Calibrated to a precision of ±0.2% of viscosity
  • Standards are NIST-traceable and certified to ASTM D2162 under ISO/IEC 17025 guidelines
PRODUCT OPTIONS
$194.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Viscosity165 cSt at 68°F; 121 cSt at 77°F; 60 cSt (281 sus) at 100°F; 54 cSt at 104°F; 35 cSt at 122°F; 7.7 cSt at 210°F; 7.5 cSt at 212°F
  • DescriptionNIST-Traceable Viscosity Standard, S60; 500 mL
Key Features
  • Standards are NIST-traceable and certified to ASTM D2162 under ISO/IEC 17025 guidelines
More About this Item
Calibrate kinematic and Saybolt viscosity equipment, both automatic and manual. A table on each bottle shows the approximate values for kinematic and Saybolt viscosity. Standards are supplied in 500-mL bottles and include full certification and a Material Data Safety Sheet.