Cole-Parmer NIST-Traceable Viscosity Standard, N10; 500 mL
Representative image only.
Cole-Parmer Item # EW-98944-70
Calibrated to a precision of ±0.2% of viscosity
  • Standards are NIST-traceable and certified to ASTM D2162 under ISO/IEC 17025 guidelines
PRODUCT OPTIONS
$282.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Viscosity21 cSt at 68°F; 17 cSt at 77°F; 11 cSt at 100°F; 10 cSt at 104°F; 7.3 cSt at 122°F; 2.7 cSt at 210°F; 2.7 cSt at 212°F
  • DescriptionNIST-Traceable Viscosity Standard, N10; 500 mL
Key Features
  • Standards are NIST-traceable and certified to ASTM D2162 under ISO/IEC 17025 guidelines
More About this Item
Calibrate kinematic and Saybolt viscosity equipment, both automatic and manual. A table on each bottle shows the approximate values for kinematic and Saybolt viscosity. Standards are supplied in 500-mL bottles and include full certification and a Material Data Safety Sheet.