Cole-Parmer High-Accuracy pH Electrode, 10-ft Cable
Representative image only.
Cole-Parmer Item # EW-27012-00
Economical and efficient combination—buy a protective housing assembly, then simply replace the electrode as needed
  • Use for many applications in a variety of housings
  • 1 year warranty
PRODUCT OPTIONS
$198.00USD / Each
BUY MORE & SAVE
  • 1-5$198.00USD / Each
  • 6-11$178.20USD / Each
  • 12-23$168.30USD / Each
  • 24+$158.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Sensing Glass TypeBlue
  • Min pH (pH)1
  • Max pH (pH)13
  • Max Temperature (° C)110
  • ConnectorBNC
  • DescriptionHigh-Accuracy pH Electrode, 10-ft Cable
  • Warranty1 year
1 Year Warranty
Key Features
  • Use for many applications in a variety of housings
  • 1 year warranty
More About this Item
High-accuracy electrodes are perfect for testing where temperature variations make many pH electrodes unreliable. These single-junction electrodes feature an innovative design that stabilizes readings in less than 20 seconds, with an accuracy of ±0.02 pH! The pH value stays constant at any given temperature up to pH 13. Maximum pressure is 100 psi at 25°>C.
Note: 100Ω RTD ATC elements are platinum and conform to DIN curve 43760. ATC connector is miniature audio plug.
Connector for the 1 kΩ RTD and the 3 kΩ Balco ATC elements is a spade lug.
Connector for 100 kΩ thermistor ATC elemen