Cole-Parmer Fluorescent Flt Yellow/Green Dye Concentrate, Pint
Representative image only.
Cole-Parmer Item # EW-00298-17
Determine water flow in closed circuits or open channels
  • Biodegradable and nontoxic
PRODUCT OPTIONS
$33.97USD / Pint of 1
$37.75USD / Pint of 1
SPECS & DESCRIPTION
Specifications & Description
  • Reagent PlatformLiquid concentrate
  • Gallons Of Water For 1 ppm (Gal)12500
  • DescriptionFluorescent Flt Yellowith Green Dye Concentrate, Pint
Key Features
  • Biodegradable and nontoxic
More About this Item
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more.
Fluorescent yellow/green dye (fluorescein) works best in silt-filled waters. Use an ultraviolet lamp (order separately below) to trace fluorescent red or yellow/green dyes where normal visual detection is difficult. Minimum detectable concentration of all dyes in clear water is 1 ppm.