Cole-Parmer Essentials Dry Clean-Wipes; 100/Canister
Representative image only.
Cole-Parmer Essentials Item # EW-33675-07
Lint-free cloth will not scratch surfaces
  • Constructed of polypropylene, wood pulp, and viscose
  • 6 x 7.5 inches
  • 1 year warranty
$14.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene, Wood Pulp, Viscose Blend
  • Width (in)6
  • Length (in)7 1/2
  • DescriptionDry Clean-Wipes; 100/Canister
  • Warranty1 year
1 Year Warranty
Key Features
  • Constructed of polypropylene, wood pulp, and viscose
  • 6 x 7.5 inches
  • 1 year warranty
More About this Item
Dry wipes can be used dry or can be used with any liquid. Simply add eight ounces of any fluid to the wipe container. Individually dispensed wipes are handy and ready for instant use. Snap-on sealed top eliminates contamination from dust or dirt. Each canister is provided in shrink-wrap or a sealed bag to maintain cleanliness for use in clean rooms. Wipes are 6" x 7.5" and packaged 100 per polyethylene canister.