Cole-Parmer Essentials Indicating Desiccant, 8 Mesh; 500g

Representative image only.
Cole-Parmer Essentials Item # EW-88401-60
Keeps samples dry and moisture free
  • CAS 7778-18-9

PRODUCT OPTIONS

$95.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeReagent
  • ClassSolids
  • FormSolid
  • Weight (g)500
  • PackagingAmber Glass Bottle
  • DOT InformationNot regulated by DOT
  • CAS Number7778-18-9
  • Shelf Life (days)60
  • DescriptionIndicating Desiccant, 8 Mesh; 500g

Key Features

  • CAS 7778-18-9

More About this Item

These desiccants are designed for use in laboratory settings and are commonly used to keep substances and samples dry, and to prevent moisture from forming on and within containers.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat