Cole-Parmer Disposable Syringe, Centric Tip, Luer Slip, 5 mL; 100/Pk

Representative image only.
Cole-Parmer Item # EW-07945-05
Ideal for all syringe filter applications
  • Constructed of durable polypropylene and polyethylene
  • Syringe features centric luer slip connection with 5 mL capacity
  • 3 month warranty

PRODUCT OPTIONS

$280.75USD / Pkg of 100

SPECS & DESCRIPTION

Specifications & Description

  • Syringe TypeCentric, Luer-Slip
  • Syringe Capacity (mL)5
  • Needle IncludedNo
  • MaterialPolypropylene and Polyethylene
  • SterileNo
  • DescriptionDisposable Syringe, Centric Tip, Luer Slip, 5 mL; 100/Pk
  • Warranty3 month
3 Month Warranty

Key Features

  • Constructed of durable polypropylene and polyethylene
  • Syringe features centric luer slip connection with 5 mL capacity
  • 3 month warranty

More About this Item

These all-plastic disposable syringes easily dispense small volumes of liquids. Polypropylene barrel and polyethylene plunger eliminate the need for rubber or synthetic gaskets. No silicone or oil lubricant is required in the syringe barrel. Choose either luer-slip or luer-lock connection type.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat