Cole-Parmer Essentials Collapsible Container, Low-Density Polyethylene (LDPE), 1 gallon; 12/PK
Representative image only.
Cole-Parmer Essentials Item # EW-06100-20
Collapsible when empty for easy storage
PRODUCT OPTIONS
$49.00USD / Pkg of 12
BUY MORE & SAVE
  • 1-5$49.00USD / Pkg of 12
  • 6-29$44.10USD / Pkg of 12
  • 30-119$41.65USD / Pkg of 12
  • 120+$39.20USD / Pkg of 12
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)4000
  • Capacity (L)4
  • Capacity (Gal)1
  • MaterialLow Density Polyethyene (LDPE)
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size38 mm
  • Cap ColorWhite
  • Bottle ColorTranslucent
  • AutoclavableNo
  • DescriptionCollapsible Container, Low-Density Polyethylene (LDPE), 1 gallon; 12/Pk
More About this Item
These containers nest uniformly when empty and expand readily when filled with liquid. They include polypropylene (PP) caps with injection-molded threads.
Replacement caps sold separately.