Cole-Parmer Clean-Wipes, 70% Alcohol/30% DI Water, ISO 6-7 Cleanroom Compatible; 100/Canister

Representative image only.
Cole-Parmer Item # EW-33675-09
Keep your surfaces clean
  • Tear-resistant and lint-free material suitable for use in ISO Class 6-7 cleanrooms
  • Presaturated in a 70% IPA and 30% water solution
  • Wipes measure 5.5 x 7 inch in dimension

$21.95USD / Pkg of 100

BUY MORE & SAVE

  • 1-23$21.95USD / Pkg of 100
  • 24-47$19.76USD / Pkg of 100
  • 48+$18.66USD / Pkg of 100

SPECS & DESCRIPTION

Specifications & Description

  • FormatPolyester & viscose
  • Width (")5 1/2
  • Height (")7
  • ApplicationsElectronics, tools, workspaces, ISO Class 6-7 cleanrooms
  • Description70% IPA Polyester/Viscose ISO 6-7 Cleanroom Wipes; 100/Canister

Key Features

  • Tear-resistant and lint-free material suitable for use in ISO Class 6-7 cleanrooms
  • Presaturated in a 70% IPA and 30% water solution
  • Wipes measure 5.5 x 7 inch in dimension

More About this Item

General-purpose, disposable 70% isopropyl alcohol (IPA) wipes are ideal for a variety of cleaning applications, including electronics, tools, workspaces, and other surfaces that require quick removal of contaminants. Durable, tear-resistant, lint-free polyester and viscose material is suitable for use in ISO Class 6-7 cleanrooms.

Wipes are presaturated in a solution of 70% isopropyl alcohol (CAS: 63-67-0) and 30% deionized water. The screw-top storage canister includes a convenient flip lid with pull-through dispenser. Each canister contains 100 perforated non-sterile wipes. Canister is foil-sealed to keep wipes moist before first use.

Available for sale in the United States only.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat