Cole-Parmer Clean-Wipes, 70% Alcohol/30% DI Water; 100/Canister
Representative image only.
Cole-Parmer Item # EW-33675-03
Strong, nonwoven cloth is soft and lint-free – will not scratch surfaces
  • No more struggling with solutions and separate towels—ready to use
  • Individually sealed containers for use in cleanrooms
PRODUCT OPTIONS
$21.50USD / Pkg of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene, Wood Pulp, Viscose Blend
  • Width (in)6
  • Length (in)7 1/2
  • DescriptionClean-Wipes, 70% Alcohol/30% DI Water; 100/Canister
Key Features
  • No more struggling with solutions and separate towels—ready to use
  • Individually sealed containers for use in cleanrooms
More About this Item
Premoistened wipes are available either with a blend of pure, reagent-grade 70% isopropyl alcohol and 30% reagent-grade deionized water, or 100% pure reagent-grade deionized water. Individually dispensed wipes are handy and ready for instant use. Snap-on sealed top eliminates contamination from dust or dirt. Each canister is provided in shrink-wrap or a sealed bag to maintain cleanliness for use in clean rooms. Wipes are 6 x 7.5 inches and packaged 100 per polyethylene canister.