Cole-Parmer Brilliant green bile broth, 500 g jar

Representative image only.
Cole-Parmer Item # EW-14200-14
Media cultures are tested and standardized to ensure consistent and reliable performance
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes

$198.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (g)500
  • DIFCO NameBrilliant Green Bile 2% (0007)
  • BBL NameBrilliant Green Bile, 2% (11080)
  • DescriptionBrilliant green bile broth, 500 g jar

Key Features

  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes

More About this Item

A standard methods medium for confirmed and completed tests for coliforms in water.

Offered in the most popular formulations, our media is thoroughly tested and subject to strict examination and standardization to ensure reliable and consistent lot-to-lot performance. Contact one of our Application Specialists to find out the availability of other dehydrated culture media and learn about our custom blending service.

Each order includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and a shelf-life guarantee. Wide mouth openings on bottles let you scoop out powder efficiently without dusting the countertop. The lightweight plastic bottles have indented hand grips for steady handling. Wide, flat cap allows you to stack containers and save space. Media are formulated to dissolve quickly.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat