Cole-Parmer Blue Tracer Dye Concentrate, 1 Pint
Representative image only.
Cole-Parmer Item # EW-00298-18
Determine water flow in closed circuits or open channels
  • Biodegradable and nontoxic
  • Blue dye brightens ponds
PRODUCT OPTIONS
$40.05USD / Pint of 1
$44.50USD / Pint of 1
SPECS & DESCRIPTION
Specifications & Description
  • Reagent PlatformLiquid concentrate
  • Gallons Of Water For 1 ppm (Gal)12500
  • DescriptionBlue Tracer Dye Concentrate, 1 Pint
NSF international certified
Key Features
  • Biodegradable and nontoxic
  • Blue dye brightens ponds
More About this Item
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more. Use blue dye to brighten murky ponds and other landscaped waters.
Minimum detectable concentration of all dyes in clear water is 1 ppm. Available in tablets, liquid, or powder. Quick dissolving tablets are easy to handling with less mess than other forms. They are perfect for applications such as storm lines, cooling and plumbing systems, and sewer lines. Choose liquid for applications such as metered input, flow/dilution/disperation rate assessment, septic inspections, or fluoremetric dye tests. Powder is well-suited for large applications where liquid is undesirable such a cooling towers, lakes, rivers, and ponds, or power plant piping.