Cole-Parmer Barometric Pressure Transmitter, 600 to 1100 hPa/mb
Representative image only.
Cole-Parmer Item # EW-98072-38
Excellent response time and long-term stability
  • Sleep mode ensures instant start-up
  • Low power consumption — 3 mA nominal
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.5 hPa/mb
  • Process Connection1/8\" hose barb
  • Battery Type9.5 to 28 VDC
  • Electrical Connections3 or 4 wire
  • Output0 to 5 VDC
  • Media CompatibilityNoncondensing gases
  • Resolution0.01 mb
  • DescriptionBarometric Pressure Transmitter, 600 to 1100 hPa/mb
  • Warranty1 year
1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Key Features
  • Sleep mode ensures instant start-up
  • Low power consumption — 3 mA nominal
  • 1 year warranty
More About this Item
Low-cost barometric pressure transducers are designed for use in environmental applications that require excellent accuracy, fast response, and long term reliability. Accurate signal conditioning and environmental compensation are provided by a capacitive sensor and proprietary custom IC circuit.
Housing is constructed of stainless steel and polyester that withstand environmental extremes. A removable 5-pin terminal strip enables easy wiring connection to datalogger and signal connections. Pressure connection is a 1/8" barbed fitting.