Cole-Parmer Alcohol Lamp with Snuffer Cap and Extra Wick, 135 mL Capacity
Representative image only.
Cole-Parmer Item # EW-36300-20
Safe and durable lamp with compact design
  • 3 year warranty
PRODUCT OPTIONS
$95.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Gas TypeAlcohol
  • Height (in)3
  • DescriptionAlcohol Lamp with Snuffer Cap and Extra Wick, 135 Ml
  • Warranty3 Years
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Alcohol lamp is ideal for laboratories where there is no gas source. It is made of nickel-plated brass. Wicks are 6" to 7" high. Optional two-way lamp stand .The distance from the lamp stand to sample is 4" or 5", depending on orientation of stand. Lamp includes snuffer cap and one extra wick.