Cole-Parmer Alcohol Lamp Two-Way Stand, Holds Samples 4 to 5" Above Lamp
Representative image only.
Cole-Parmer Item # EW-36300-26
Safe and durable lamp with compact design
PRODUCT OPTIONS
$35.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Length (in)4
  • Height (in)5
  • DescriptionAlcohol Lamp Two-Way Stand, Holds Samples 4 to 5\" Above Lamp
More About this Item
Alcohol lamp is ideal for laboratories where there is no gas source. It is made of nickel-plated brass. Wicks are 6" to 7" high. Optional two-way lamp stand .The distance from the lamp stand to sample is 4" or 5", depending on orientation of stand. Lamp includes snuffer cap and one extra wick.