Cole-Parmer Adapter, U.S. standard meter to BNC electrode
Representative image only.
Cole-Parmer Item # EW-05983-25
  • 6 month warranty
PRODUCT OPTIONS
$72.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionAdapter, U.S. standard meter to BNC electrode
  • Warranty6 month
6 Month Warranty
Key Features
  • 6 month warranty
More About this Item
BNC is the most common electrode connector, but some pH meters and electrodes come with different types of connectors. Order an adapter to ensure your meter and electrode are compatible.