Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Transmitter

Representative image only.
Cole-Parmer Item # EW-05802-14
Automatic buffer recognition for quicker setup
  • Backlight LCD for clear measurement display
  • Automatic buffer recognition of US and NIST buffers
  • Universal power supply input from 100 to 240VAC
  • Easy-to-install 1/8-DIN compact plastic casing
  • Isolated 4 to 20 mA current output allows you to open valves, pace pumps, or transmit your measurment to external equiptment
  • 1 year warranty

PRODUCT OPTIONS

$293.00USD / Each

$164.50 - $237.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Often Bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Control TypeOn/Off
  • Display TypeLCD
  • Housing Material1/8-DIN
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01
  • pH Accuracy±2% of reading
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution1mV
  • mV Accuracy±2mV
  • Min Temperature (° C)-10
  • Max Temperature (° C)120
  • Min Temperature (° F)14
  • Max Temperature (° F)248
  • Temperature Resolution0.1°
  • Temperature Accuracy±0.5°C
  • Temperature CompensationAutomatic
  • Output Current4-20mA
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • Width (cm)9.6
  • Height (cm)4.8
  • Depth (cm)11
  • Width (in)3 21/64
  • Height (in)1 3/4
  • Depth (in)4 1/4
  • Panel Mount Size1/8-DIN
  • Description350 pH/ORP/Temperature 1/8-DIN Transmitter; with Isolated 4 to 20 mA output
  • Warranty1 year
1 Year WarrantyConformite Europeenne

Key Features

  • Backlight LCD for clear measurement display
  • Automatic buffer recognition of US and NIST buffers
  • Universal power supply input from 100 to 240VAC
  • Easy-to-install 1/8-DIN compact plastic casing
  • Isolated 4 to 20 mA current output allows you to open valves, pace pumps, or transmit your measurment to external equiptment
  • 1 year warranty

More About this Item

Cole-Parmer pH/ORP controllers offer features found in more expensive models. Automatic buffer recognition, absolute and relative ORP measurements, and automatic temperature compensation ensure measurement accuracy. Models with two sets of programmable on/off relays are ideal for high/low control to activate an alarm, a pump, or another device. Models with an isolated 4-20 mA output signal are suitable for opening valves and pacing pumps. ATC element is user selectable, 10K NTC or 1000 Ohm RTD. Order 56105-60 BNC-to-Stripped Wire Adapter for easy connections on terminal strip on back of controller.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat