Cole-Parmer 1 To 2000 mTorr Pressure/vacuum Pirani-Type Sensor, 1/8" NPT(M)
Representative image only.
Cole-Parmer Item # EW-68801-51
Customize to your needs
  • Complete system requires:
  • 1. Sensor
  • 2. Indicator or controller
  • Optional 3. Calibrator
$171.75USD / Each
$204.00 - $277.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±5% of reading or ±3 mTorr
  • Wetted Materials304 SS
  • Process Connection1/8\" NPT(M)
  • Media CompatibilityGases compatible with wetted materials
  • Resolution1 mTorr
  • Description1 To 2000 Mtorr Pressure/vacuum Pirani-type Sensor, 1/8\" NPT(M)
Key Features
  • Complete system requires:
  • 1. Sensor
  • 2. Indicator or controller
  • Optional 3. Calibrator
More About this Item
Sensors provide improved tracking ability and maintain calibration over long periods of time. When replacing a sensor, you do not need to recalibrate.
These sensors are suited for use with mercury and oil pumps.
Pirani type sensors offer enhanced stability and an automatic temperature compensation network. The 304 stainless steel sensors operate at half the
temperature of standard thermopile sensors. Use for high-vacuum applications.
Diaphragm type sensor offers quick response—less than 400 milliseconds full-scale. Select a diaphragm type sensor made from 316 stainless steel or silicon based on your application.