CELLTREAT Scientific Products 230200 Electronic Pipette Controller; 120 VAC

Representative image only.
CELLTREAT Scientific Products Mfr # 230200 – Item # EW-25200-07
Take wherever needed in the lab—cordless and rechargeable
  • Adjust speed with the turn of a dial
  • Obtain up to 2500 pipettings (up to 8 hours of use) on a single charge

$353.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • SterileNo
  • Min Volume (mL)1
  • Max Volume (mL)100
  • Number Of Channels1
  • SpeedAdjustable
  • Power (VAC)120
  • BatteryRechargeable
  • DescriptionElectronic Pipette Controller; 120 VAC

Includes

adhesive mounting bracket, rubber grommet/nosepiece, 0.45-µm pore filter, rechargeable battery, and 120 VAC power adapter/charger.

Key Features

  • Adjust speed with the turn of a dial
  • Obtain up to 2500 pipettings (up to 8 hours of use) on a single charge

More About this Item

This CELLTREAT pipette controller adapts to fill and dispense 1- to 100-mL sized pipettes. Control speed with the turn of a dial, and direction with low-pressure buttons. Avoid accidental over aspiration, contamination, and damage to the controller from volatile vapors with the included filter.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat