CELLTREAT Scientific Products 229584 Non-Treated Culture Roller Bottle, 2L, nonvented cap, sterile, 12/cs
Representative image only.
CELLTREAT Scientific Products Mfr # 229584 – Item # EW-01836-38
Uniform, hydrophilic surface promotes cell attachment
  • Uniform, hydrophilic surface for cell attachment
  • Nontreated roller bottles
  • Solid, nonvented caps
PRODUCT OPTIONS
$107.50USD / Case of 12
Often bought with…
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolystyrene
  • SterileYes
  • Capacity (mL)2000
  • DescriptionNon-Treated Culture Roller Bottle, 2L, nonvented cap, sterile, 12/cs
Sterile
Key Features
  • Uniform, hydrophilic surface for cell attachment
  • Nontreated roller bottles
  • Solid, nonvented caps
More About this Item
Bottles are made of clear polystyrene and feature easy-to-read black graduations. Each bottle is gamma irradiated and individually bagged. Caps have large knurls for easy opening.