BRAND Dispensette® S Organic Bottletop Dispenser, Digital Adjustable, with Recirculation Valve, 1-10 mL

Representative image only.
Mfr # 4630341 – Item # EW-07915-62
Best choice for concentrated acids and organic solvents
  • Proven floating piston design provides one handed operation even for large volumes
  • Numerous safety features
  • Chemically resistant materials, such as PTFE, ETFE, PFA, FEP, Al203, platinum-iridium and borosilicate glass
  • Hinged screw cap hinges away from dispensing when removed
  • Available with priming valve
  • 1 year warranty

PRODUCT OPTIONS

$767.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Min Volume (mL)1
  • Max Volume (mL)10
  • Division (mL)0.05
  • Accuracy±0.5%
  • Reproducibility≤ 0.1%
  • AutoclavableYes
  • Wetted Partsborosilicate glass, aluminum oxide ceramic, tantalum, ETFE, FEP, PFA, PTFE, and PP
  • DescriptionOrganic Dispenser, 1 - 10 mL, digital, recirculating
  • Warranty1 year
1 Year Warranty

Key Features

  • Proven floating piston design provides one handed operation even for large volumes
  • Numerous safety features
  • Chemically resistant materials, such as PTFE, ETFE, PFA, FEP, Al203, platinum-iridium and borosilicate glass
  • Hinged screw cap hinges away from dispensing when removed
  • Available with priming valve
  • 1 year warranty

More About this Item

This fourth-generation Dispensette® S Organic bottletop dispenser is an excellent choice for general dispensing. A reinforced discharge tube with swing-away screw cap improves safety while an improved fluid path requires even lower operating forces. A new safety discharge valve reduces the chance of dispensing unless the discharge tube is mounted properly.
Choose a dispenser with a recirculation valve if you want chemicals to route back into the reagent bottle during the priming step. Analog models feature adjustable volume selection with interior scalloped track to provide reproducibe settings.
Dispensette® S Organic Dispensers are optimized for use with concentrated acids and organic solvents. Not for use with hydrofluoric acid.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat