Bradley S19-220 Eyewash Bowls, Wall Mount, ABS Plastic
Representative image only.
Bradley Mfr # S19-220 – Item # EW-86000-60
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Eyewash Fixture MaterialChrome-plated brass
  • Bowl MaterialABS plastic
  • MountWall Mount
  • Outlet Connections1-1/4\" NPT(F)
  • Inlet Connection1/2\" NPT(F)
  • ComplianceANSI Z358.1-2009
  • DescriptionEyewash Bowls, Wall Mount, ABS Plastic
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
All bowls are 10" (25 cm) in diameter and come with dual sprayhead assemblies and PVC push handles to start flow.
Wall-mount model includes brackets and comes in yellow impact-resistant ABS plastic (drain fittings and trap not included).
Compliance: ANSI Z358.1-2004