Big Blue 155120-43 Filter cartridge, "", 10", pleated polypropylene, 30 µm
Representative image only.
Big Blue Mfr # 155120-43 – Item # EW-01508-86
Design offers higher flow rates and dirt-loading capacities than spun or wound depth cartridges, especially when removing lower micron-sized particles
  • Remove non-gelatinous particles that fall into a narrow size range, typically in post filtration or polishing applications
PRODUCT OPTIONS
$32.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Filter MediaPolypropylene
  • Micron Rating30
  • Filter Length (in)10
  • DescriptionFilter cartridge, "", 10", pleated polypropylene, 30 µm
Key Features
  • Remove non-gelatinous particles that fall into a narrow size range, typically in post filtration or polishing applications
More About this Item
Pleated cartridges separate viscous sediments and flat particulates. Use polyester cartridges for water filtration and polypropylene cartridges with harsher chemicals. Plastisol end caps.
Note: Cartridges are for sediment filtration only; they will not remove bacteria.