Bel-Art Poxygrid Benchtop Biohazard Bag Stand
Representative image only.
Bel-Art Mfr # H13193-1000 – Item # EW-06496-60
Comply with OSHA standards for biohazardous material handling
  • Epoxy-coated steel wire stand is durable and easy to clean
  • All components are autoclavable
PRODUCT OPTIONS
$56.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ColorOrange
  • MaterialEpoxy-coated steel wire
  • Length (in)11
  • Width (in)8.5
  • Length (cm)28
  • Width (cm)22
  • DescriptionPoxygrid Benchtop Biohazard Bag Stand
Key Features
  • Epoxy-coated steel wire stand is durable and easy to clean
  • All components are autoclavable
More About this Item
These orange colored epoxy-coated steel wire stands can securely hold a high-density 8 1/2" x 11" (22 x 28 cm) polyethylene bag in an upright open position. Ideal for collecting pipette tips, microcentrifuge tubes, and other hazardous materials throughout the day. The stands are reusable, can be cleaned by hand or in automated dish washer, and autoclavable at 250°F (121°C).