B&K Precision 601 Battery Capacity Analyzer 6V & 12V SLA
Representative image only.
B&K Precision Mfr # 601B – Item # EW-20020-75
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DC Min (Volts)6
  • DC Max (Volts)12
  • AC Min (Volts)6
  • AC Max (Volts)12
  • DescriptionBattery Capacity Analyzer 6V & 12V SLA
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This lightweight, hand-held battery capacity analyzer provides quick, accurate test of percentage capacity in 12V lead acid batteries. Its small
size and easy portability make it ideal for field and shop service and maintenance applications with automotive fleets, communication equipment, UPS manufacturing and maintenance, and backup lighting systems.
FEATURES
  • Measures both no load voltage and battery capacity
  • Displays storage capacity of 12V lead acid batteries as a percentage
  • Great tool for testing back up batteries for UPS, security, emergency flood light systems
  • Ideal for automotive, UPS maintenance and telecommunication applications
  • Powered by battery under test