Aven Tools 26700-207 Digital Near-Infrared Microscope, 1.3 megapixels, 10x to 200x; USB
Representative image only.
Aven Tools Mfr # 26700-207 – Item # EW-48708-26
Puts the world of microphotography at the tips of your fingers!
  • Active Pixel Technology produces sharp images and accurate color reproduction
  • Six built-in lights with adjustable brightness ensure proper illumination
  • Includes software for image/video capture and measurement
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification10x to 200x
  • Maximum Magnification200x
  • Resolution1,300,000 pixels
  • Illuminator(s)Six near-infrared LEDs
  • Image Sensor1/2\" CMOS
  • OutputUSB
  • Cable(s)USB 2.0
  • Power5 VDC, via USB
  • DescriptionDigital Near-Infrared Microscope, 1.3 megapixels, 10x to 200x; USB
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • Active Pixel Technology produces sharp images and accurate color reproduction
  • Six built-in lights with adjustable brightness ensure proper illumination
  • Includes software for image/video capture and measurement
  • 1 year warranty
More About this Item
Use these digital USB microscopes for science and engineering work, assembly and quality control, detailed repair, forensics, and more. Six LEDs illuminate your object — turn the LEDs on or off and adjust the brightness using the control wheel on the body. For versions with the 10x to 200x magnification range, magnification automatically adjusts based on the distance from the object. Order optional boom stand for greater flexibility.
Capture images and videos through use of the included software (requires Windows® XP, SP2, or Vista). The software also includes a measurement function, brightness and contrast controls, digital zoom, and date and time record.