Ashcroft IXLDP3IWC Low Diff Pressure Transmitter, 3.0" WC, 4-20mA, 0.5%

Representative image only.
Ashcroft Mfr # IX-5-F02-42-ST-3IW – Item # EW-68439-41
  • FEATURES
  • Certified 0.25% and 0.50% accuracy
  • High over pressure protection
  • NEMA 4X metal construction
  • Six output signals
  • Easy installation
  • FM approved for hazardous locations (optional)
  • 1 year warranty
  • SPECS & DESCRIPTION

    Specifications & Description

    • Accuracy±0.5% full-scale
    • Min Pressure (in WC)0
    • Max Pressure (in WC)3
    • Min Temperature (° F)-20
    • Max Temperature (° F)185
    • Min Temperature (° C)-29
    • Max Temperature (° C)85
    • Wetted Materials300 SS, silicone/glass
    • Case300 SS
    • Process Connection1/4\" NPT(F)
    • Unit Of Measurement\" WC
    • Battery Type12-36 VDC
    • Electrical Connections1/2\" NPT(F) conduits
    • Electrical terminationScrew terminals
    • Output4 to 20 mA
    • Enclosure RatingNEMA 4X
    • Media CompatibilityClean, dry, noncorrosive gases
    • DescriptionLow Diff Pressure Transmitter, 3.0\" WC, 4-20mA, 0.5%
    • Warranty1 year
    1 Year Warranty

    Key Features

    • FEATURES
    • Certified 0.25% and 0.50% accuracy
    • High over pressure protection
    • NEMA 4X metal construction
    • Six output signals
    • Easy installation
    • FM approved for hazardous locations (optional)
  • 1 year warranty
  • More About this Item

    Designed for the measurement and control of very low pressure and flow in industrial and process plant environments. The transmitter features s rugged NEMA4X enclosure, built-in electrical terminal box isolated from the electronics and threaded process connections.

    REVIEWS

    Q&A

    CUSTOMER SUPPORT

    Need Help with this product?

    Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

    Call Us At1-800-323-4340
    Live chat