Ashcroft 14902.5 Dual-Scale Vacuum Gauge 15" WC and 4 kPa
Representative image only.
Ashcroft Mfr # 25-1490-A-02L-XTU-15IWV/KPA – Item # EW-68930-30
Effective low-pressure and low-vacuum response with diaphragm
  • True zero indication identifies damaged gauges increasing safety and ensuring accuracy
  • Built-in snubber minimizes rapid pressure fluctuations
  • 1 year warranty
PRODUCT OPTIONS
$59.20USD / Each
$74.00USD / Each
$152.00 - $225.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±2-1-2% full-scale (ANSI/ASME Grade A)
  • Dial Size (in)2.5
  • Dial Size (mm)64
  • Min Pressure (in WC)-15
  • Max Vacuum (\" Hg)1.1
  • Wetted MaterialsBeryllium copper, brass, polysulfone, RTV silicone
  • CasePolysulphone
  • Lens MaterialPolycarbonate
  • Process Connection1/4\" NPT(M), lower
  • Unit Of Measurement\" WC, kPa
  • Displayanalog dial
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionDual-Scale Vacuum Gauge 15\" WC and 4 kPa
  • Warranty1 year
1 Year Warranty
Key Features
  • True zero indication identifies damaged gauges increasing safety and ensuring accuracy
  • Built-in snubber minimizes rapid pressure fluctuations
  • 1 year warranty
More About this Item
Gauges have brass socket, beryllium copper diaphragm capsule, painted steel case, and polycarbonate lens. Bottom connection is 1/4" NPT(M).