Ashcroft 11302.5 2-½" Economical Differential Pressure Gauges, 0 to 30 psid
Representative image only.
Ashcroft Mfr # 25-1130-FD-25S-30# – Item # EW-68403-04
Excellent for filtration systems, leak monitoring, and maintaining flow and tank levels
  • Close-coupled magnetic rack and pinion design achieves superior stability.
  • Weather-resistant stainless steel case, aluminum body, and Buna N seals
  • Use glycerin filled gauges in high vibration areas to achieve reliable and stable readings.
  • TrueZero™ indication for dependable pressure readings at all times within ±2% full-scale accuracy (ascending)
  • 5 year warranty
PRODUCT OPTIONS
$148.00USD / Each
$185.00USD / Each
$152.00 - $225.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±2% full-scale (ascending)
  • Dial Size (in)2.5
  • Dial Size (mm)64
  • Min Pressure (PSID)0
  • Max Pressure (PSID)30
  • Min Temperature (° F)-50
  • Max Temperature (° F)175
  • Min Temperature (° C)-45
  • Max Temperature (° C)79
  • Wetted Materials316L SS
  • CaseStainless Steel
  • Lens MaterialGlass
  • Process Connection1/4\" NPT(F), in-line
  • Unit Of Measurementpsid
  • DisplayAnalog dial
  • Media CompatibilityGases and liquids compatible with wetted materials
  • Description2-½\" Economical Differential Pressure Gauges, 0 to 30 psid
  • Warranty5 years
5 Year Warranty
Key Features
  • Close-coupled magnetic rack and pinion design achieves superior stability.
  • Weather-resistant stainless steel case, aluminum body, and Buna N seals
  • Use glycerin filled gauges in high vibration areas to achieve reliable and stable readings.
  • TrueZero™ indication for dependable pressure readings at all times within ±2% full-scale accuracy (ascending)
  • 5 year warranty