Ashcroft 11254.5 Gauge with 0 to 30 psid Range, Bottom Connection
Representative image only.
Ashcroft Mfr # 45-1125-02L-30# – Item # EW-68402-02
  • 1 year warranty
$1,138.00USD / Each
$152.00 - $225.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±2-1-2% (ANSI/ASME Grade A)
  • Dial Size (in)4.5
  • Dial Size (mm)114
  • Min Pressure (PSID)0
  • Max Pressure (PSID)30
  • Process Connection1/4\" lower
  • Unit Of Measurementpsid
  • Resolution0.5
  • DescriptionGauge with 0 to 30 psid range, bottom connection
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This differential gauge features ASME B40.1 grade 1A accuracy, bronze Bourdon tube, and black epoxy-coated aluminum case.