Cole-Parmer Essentials Cell and Tissue Culture Plates with Lid, Sterile, Polystyrene, Treated, 96-Well; 100/CS

Representative image only.
Cole-Parmer Essentials Item # EW-01927-78
Extremely smooth surface and raised well rims reduce evaporation and maximize results
  • Gamma sterilized and nonpyrogenic
  • Individually packaged in peel-to-open blister pack
  • Printed lot numbers on both plate and packaging for effortless traceability
  • Optically clear plates for easy visualization of cell growth
  • Features treated surface that provides a hydrophilic surface ideal for cell attachment
  • Assembled plate and lid height: 16.0 mm
  • Max volume (single well): 0.39 mL
  • Working volume (single well): 0.075 to 0.2 mL
  • Growth area (single well): 0.33 cm²

PRODUCT OPTIONS

$170.00USD / Case of 100

SPECS & DESCRIPTION

Specifications & Description

  • Number of Wells96
  • Min Working Well Volume (mL)0.075
  • Max Working Well Volume (mL)0.2
  • Well ShapeFlat
  • Culture Area (cm²)0.33
  • Surface TreatmentYes
  • Lid (Included)Yes
  • SterileYes
  • DescriptionCell and Tissue Culture Plates with Lid, Sterile, Polystyrene, Treated, 96-Well; 100/CS
NonpyrogenicSterile

Includes

Lids

Key Features

  • Gamma sterilized and nonpyrogenic
  • Individually packaged in peel-to-open blister pack
  • Printed lot numbers on both plate and packaging for effortless traceability
  • Optically clear plates for easy visualization of cell growth
  • Features treated surface that provides a hydrophilic surface ideal for cell attachment
  • Assembled plate and lid height: 16.0 mm
  • Max volume (single well): 0.39 mL
  • Working volume (single well): 0.075 to 0.2 mL
  • Growth area (single well): 0.33 cm²

More About this Item

Use these sterile well plates to serve your various cell and tissue culture needs. With individual condensation rings and a one-way, tight-fitting lid, they reduce cross-contamination and help prevent evaporation. An alphanumeric notation on each well aids in simple identification. Transparent polystyrene construction allows for easy visualization of cell growth and facilitates observation under a microscope.

Manufactured under ISO 13485 and ISO 9001 quality management systems in an ISO Class 8 (formerly Class 100,000) cleanroom environment for optimal quality, these cell and tissue culture well plates are individually packaged in peel-to-open blister packs, sterilized by irradiation (SAL 10-6), and are DNase/RNase free, nonpyrogenic, and noncytotoxic. They conform to USP Class VI and are quality traceable with the lot number on both the plate and packaging.

Plates with a treated surface are suitable for common adherent cell lines due to the excellent hydrophilicity of the surface while untreated plates are suitable for suspension cell culture. Compatible with most multi-well plate instruments and equipment.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat