Argos Technologies PolarSafe® 2D Cryogenic Vials, Sterile, Skirted-Bottom, Internal Thread, 5 mL; 50/Pk

Representative image only.
Argos Technologies Item # EW-04395-83
Embedded Datamatrix code provides fast and accurate sample processing
  • Innovative “channels of evacuation” design allows fluid to easily drain from the vials

PRODUCT OPTIONS

$126.00USD / Pkg of 50
Include Barcode Scanner Today

SPECS & DESCRIPTION

Specifications & Description

  • Volume (mL)5
  • Tube MaterialPolypropylene
  • Tube Bottom ShapeSkirted
  • Cap TypeScrew cap
  • Cap MaterialPolyethylene and rubber
  • ThreadInternal
  • SterileYes
  • AutoclavableYes
  • Height (mm)89
  • Outside Diameter (mm)12.6
  • Description2D Cryogenic Vials, Sterile, Skirted-Bottom, Internal Thread, 5 mL; 50/Pk

Key Features

  • Innovative “channels of evacuation” design allows fluid to easily drain from the vials

More About this Item

The PolarSafe cryogenic 2D vials, available in 2 and 5 mL sizes with internally or externally threaded caps, are idea for rapid processing of biological samples. The embedded Datamatrix code at the bottom of each vial along with the corresponding “Code 128” barcode on the body of the vial, saves time and provides accurate organization and tracking of valuable samples.

The 2D vials can be safely stored in polycarbonate boxes (purchase separately). The storage boxes also feature the 2D Datamatrix code and Code 128 barcode to provide automatic identification of both the box and stored vials.


Rated for use down to -196°C. Liquid phase storage may cause entrapment of liquefied nitrogen inside the vial and lead to pressure build-up, resulting in possible explosion or biohazard release. It is recommended storing in the gas phase above the nitrogen (-178 to –150ºC) to prevent issue. Recommend filling the tube with no more than 90% of the total nominal volume to permit sample expansion.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat