Argos Technologies Pipette Tip Box, Polycarbonate, 200ul, 128-Place Grid Insert; Yellow Rack
Representative image only.
Argos Technologies Item # EW-24406-65
Ideal for pipette tip protection and storage
  • Transparent box contains a hinged lid for easy one hand opening
  • Autoclavable at 121°C
  • Constructed of durable polycarbonate
  • Accommodates 200 μl pipette tips
  • Features a 128-place grid insert for quick organization
  • Dimensions of 6-3/4" L x 3-3/16" H x 3-9/16" W
  • Designed with transparent box and yellow pipette rack
SPECS & DESCRIPTION
Specifications & Description
  • Width (InchShort)3 9/16
  • AutoclavableYes
  • Height (in)3 3/16
  • Overall Length (in)6.75
  • Wetted MatterialPolycarbonate
  • Color CodeYellow
  • Compatible with128 x 200 ul Pipette Tip
  • DescriptionPipette Tip Box, Polycarbonate, 200ul, 128-Place Grid Insert; Yellow Rack
Key Features
  • Transparent box contains a hinged lid for easy one hand opening
  • Autoclavable at 121°C
  • Constructed of durable polycarbonate
  • Accommodates 200 μl pipette tips
  • Features a 128-place grid insert for quick organization
  • Dimensions of 6-3/4" L x 3-3/16" H x 3-9/16" W
  • Designed with transparent box and yellow pipette rack
More About this Item
Choose from pipette storage boxes with included pipette tip rack or storage box with removable divider.