Cole-Parmer Essentials Reagent Reservoir, Sterile, 1-Channel, 50 mL, Individually Wrapped; 50/Cs
Representative image only.
Cole-Parmer Essentials Item # EW-04395-26
V-shaped angled design maximizes reagent retrieval and reduces waste
  • Certified RNase-, DNase-, and pyrogen-free
PRODUCT OPTIONS
$45.00USD / Case of 50
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Max Volume (mL)50
  • Number Of Channels1
  • Wetted MatterialPolystyrene
  • Color CodeWhite
  • Package OfIndividually Wrapped 50/Case
  • Compatible with8- and 12-Channel Pipettors
  • DescriptionReagent Reservoir, Sterile, 1-Channel, 50 mL, Individually Wrapped; 50/Cs
Key Features
  • Certified RNase-, DNase-, and pyrogen-free
More About this Item
The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents.
Single Chamber Reservoirs are made of white polystyrene and constructed with low cross bars at each end for added stiffness and strength. Each reservoir features graduations on the inner walls, pour spouts in every corner, and one larger spout to facilitate spill-free pouring. Reservoirs are ideal for 8- and 12-channel pipettors.
12-Chamber Reservoirs are designed for serial dilutions or pipetting different reagents using a 12-channel pipettor. Each numbered chamber holds up to 3 mL. The chemical-resistant polypropylene reservoir can withstand temperatures from -122 to 250°F (-80 to 121°C). Tight fitting lid minimizes evaporation or contamination during incubation or storage.