Cole-Parmer Essentials OmegaZen™ Power Supply for As One; Japan
Representative image only.
Cole-Parmer Essentials Item # EW-25303-99
Enhance and maintain your Omega Zen pipettors with these accessories
  • Compatible with Omega Zen pipette controllers
  • Choose from a wide range of accessories to make your pipettor even more versatile
  • Power supply compatible with Omega Zen pipettor for use in Japan
  • 1 year warranty
PRODUCT OPTIONS
$45.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)100 to 240
  • Compatible withArgos Technologies OmegaZen Pipette Controller
  • DescriptionPower Supply for As One; Japan
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Compatible with Omega Zen pipette controllers
  • Choose from a wide range of accessories to make your pipettor even more versatile
  • Power supply compatible with Omega Zen pipettor for use in Japan
  • 1 year warranty
More About this Item
These accessories can be used to enhance the flexibility of your Omega Zen controller, or be used for routine maintenance. There are four filter options to choose from. All are made of a polypropylene housing with a hydrophobic PTFE membrane. The Omega Zen controllers accept either the 0.2 µm or the 0.45 µm pore size filters.
Power your Omega Zen controller with the rechargeable lithium-ion battery for up to 9 hours of continuous use and charge the battery with the universal power supply.
The nosepiece and silicone pipette adapter are both autoclavable for optional sterility.