Applikon BioBundle Z001059051 pH Sensor for Fermentation System, 590 mm
Representative image only.
Applikon BioBundle Mfr # Z001059051 – Item # EW-29207-28
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionpH Sensor for Fermentation System, 590 mm
More About this Item
These pH sensors use a nontoxic gel to prevent silver/protein interference on the diaphragm. Each sensor has a Ti layer on the innertube to ensure no window between sensor head and gel which results in an excellent screen to protect against electromagnetic interference. Pre-autoclaved; MSDS and GLP certificate available upon request.