Cole-Parmer Essentials Anti-Static Weighing Funnel, PP, Medium; 50/PK
Representative image only.
Cole-Parmer Essentials Item # EW-06127-04
Ensure complete sample transfer
  • Autoclavable
  • 6 month warranty
  • 6 month warranty
PRODUCT OPTIONS
$114.00USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene (PP)
  • Stem OD (mm)10.35
  • Overall Length (mm)61.02
  • Overall Length (in)2 2/5
  • Fits Flask Size100 to 250 mL
  • DescriptionAnti-Static Weighing Funnel, PP, Medium; 50/PK
  • Warranty6 month
6 Month Warranty
Key Features
  • Autoclavable
  • 6 month warranty
  • 6 month warranty
More About this Item
These disposable polypropylene (PP) weighing funnels are designed to fit in volumetric flasks. One side of the funnel is flattened so you can set the funnel flat on a balance.
Certificate of Analysis
For a sample Certificate of Analysis, please go to link below under RESOURCES.
If a Lot specific Certificate of Analysis is required, please call 1-800-323-4340 for assistance.