Amprobe AF-600 A-Frame Fault Locator, for use with UAT-600 Series
Representative image only.
Amprobe Mfr # Amprobe AF-600 – Item # EW-20034-75
Saves money by pinpointing leakage point around a cable
  • Optional accessory for Amprobe UAT-600 Underground Utilities Locator Series
  • Works in conjunction with the UAT-600-T Transmitter
  • Locate cable and wires ground faults, sheath faults or pipeline coating defects (direct contact of the utility with the ground is required to pinpoint a fault)
  • Used to find the point where metal is touching the ground (shield is rusted/rubber buffer is broken) and power is leaking, creating noise on a cable
  • Adjustable volume control
  • Advanced technology and digital signal processing makes pinpointing process fast, accurate and unambiguous
  • 1 year warranty
$3,021.99USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • Current Frequency8 kHz
  • Display1.28 in, 128 x 128 BW outdoor LCD display with auto backlight
  • DescriptionAF-600 A-Frame Fault Locator, for use with UAT-600 Series
  • Warranty1 Year
1 Year Warranty
Key Features
  • Optional accessory for Amprobe UAT-600 Underground Utilities Locator Series
  • Works in conjunction with the UAT-600-T Transmitter
  • Locate cable and wires ground faults, sheath faults or pipeline coating defects (direct contact of the utility with the ground is required to pinpoint a fault)
  • Used to find the point where metal is touching the ground (shield is rusted/rubber buffer is broken) and power is leaking, creating noise on a cable
  • Adjustable volume control
  • Advanced technology and digital signal processing makes pinpointing process fast, accurate and unambiguous
  • 1 year warranty
More About this Item
The AF-600 A-Frame cable ground fault finder is specifically designed for the Amprobe UAT-600 Underground Utilities Locator Series. In combination with the UAT-600 Transmitter, it will pinpoint the place where a cable metal conductor (either a sheath or a metallic conductor of the wire) touches the ground. It can also detect other conductors to ground faults such as pipeline coating defects.