Advantec USY-20 Disposable Ultrafiltration Unit, 200K MWCO, pack of 24

Representative image only.
Advantec Mfr # USY-20 – Item # EW-02910-54
Concentrate, separate, and purify your biological samples without a centrifuge
  • Three molecular weight cutoffs
  • 40 times maximum concentration
  • 1 year warranty

PRODUCT OPTIONS

$175.50USD / Pkg of 24

SPECS & DESCRIPTION

Specifications & Description

  • Hold-Up Volume (mL)0.02
  • Max Temperature (° C)50
  • Max Pressure (bar)2.9
  • Maxi Pressure (Mpa)0.29
  • Nominal Molecular Weight Limit (Mwl) (Dalton)200000
  • Filtration Area (cm²)2.0
  • Max Molecular Weight Cut Off (Mwco) (Dalton)200000
  • Max Pressure (PSI)43
  • Max Temperature (° F)122
  • Min Volume Range (mL)0.02
  • Max Volume Range (mL)2
  • DescriptionDisposable Ultrafiltration Unit, 200K MWCO, pack of 24
  • Warranty1 year
1 Year Warranty

Key Features

  • Three molecular weight cutoffs
  • 40 times maximum concentration
  • 1 year warranty

More About this Item

These disposable ultrafiltration units are designed for ease in concentrating, separating, or otherwise purifying small volumes (<2 mL) of liquid samples, on the basis of molecular weight cutoff using positive pressure. The unit is constructed of polypropylene cap, acrylic retentive cell, and filtrate cell. Filter media is polysulfone. To use, simply dispense sample into the retentive cell by pipette, place cap on retentive cell, and inject 10 mL of air with a syringe. Remove cap to stop filtration. Sterilization: 25% ethanol, 5% formalin.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat