Advantec GA100125MM Grade GA100 Glass Fiber Filters; 125mm dia; 100/box
Representative image only.
Advantec Mfr # GA100125MM – Item # EW-06657-56
For analytical or gravimetric studies
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass Microfiber
  • Diameter (mm)125
  • Diameter (cm)12.5
  • DescriptionGrade GA100 Glass Fiber Filters; 125mm dia; 100/box
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
General purpose; filtering cells and precipitated proteins; air pollution monitoring.