Additel ADT100-HTK-NPTM4-NPTF3Q Pressure Hose; 1/4"NPTM, 3/8"NPTF Quick-Connect
Representative image only.
Additel Mfr # ADT100-HTK-NPTM4-NPTF3Q – Item # EW-16108-22
Convenient for fast and tool-free calibrations
PRODUCT OPTIONS
$245.00USD / Each
sign in for your price.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPressure Hose; 1/4"NPTM, 3/8"NPTF Quick-Connect
More About this Item
Each component is precision machined so that leak-free connections can be made by hand—no tools or tape necessary!
Hose test kit includes 5 ft (1.5 m) flexible hose (15,000 psi) and 1/4" NPT(M) to 3/8" NPT(F) hand-tight quick connector.