Adam Equipment 1070010636 Temperature Calibration Kit for PMB Moisture Analyzer
Representative image only.
Adam Equipment Mfr # 1070010636 – Item # EW-11120-80
Ensure accuracy of your balance
  • Simple to use with special grip to position temperature sensor in the right place
  • Large display of temperature reading
$227.50USD / Each
$260.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeExternal
  • RS-232 InterfaceYes
  • Auto ShutoffY
  • Low Battery IndicatorYes
  • Battery1 LR44 1.5V Button Cell (included)
  • DescriptionTemperature Calibration Kit for Pmb Moisture Analyzer
Includes
Carrying case, device holder, and LR44 button cell battery.
Key Features
  • Simple to use with special grip to position temperature sensor in the right place
  • Large display of temperature reading
More About this Item
Moisture Analyzers should be calibrated frequently, just like balances. Calibrate the temperature sensor of your PMB Moisture Analyzer with this Temperature Calibration Kit, which comes with a case and a device to hold the sensor in the optimum position.